missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promed Bioscience Atelocollagen, Porcine Type I, Ultrapure (Enzymatic extraction, dialyzed)
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
Description
- Lyophilized, ultrapurified (via dialysis) Type I atelocollagen with superior functionality and physical/mechanical properties (strength, elasticity)
- The proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
- Suitable as bioink for 3D bioprinting, for in vitro cell cultures, for cell functional assays and for in vitro and in vivo research applications (wound healing, tissue regeneration, drug delivery)
Specifications
Specifications
| Formulation | Lyophilized Fibrous Powder |
| Purification Method | Double Dialysis |
| Quantity | 20 mg |
| Content And Storage | -20°C to 8°C |
| Endotoxin Level | <1.0 EU/mL |
| Purity or Quality Grade | >97% (by SDS/Western blotting) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction