missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HACE1 Monoclonal Antibody (OTI6H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810071S
This item is not returnable.
View return policy
Description
HACE1 Monoclonal antibody specifically detects HACE1 in Human samples. It is validated for Western BlotSpecifications
| HACE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZ95 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54904 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI6H8 | |
| Unconjugated | |
| Nsd3 | |
| 1700042J16Rik; A730034A22Rik; BC025474; E3 ubiquitin-protein ligase HACE1; hace 1; Hace1; HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1; HECT-type E3 ubiquitin transferase HACE1; KIAA1320; SPPRS | |
| Human recombinant protein fragment corresponding to amino acids 427-566 of human WHSC1L1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction