Learn More
HACE1 Mouse anti-Human, Clone: OTI6H8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810071
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
E3 ubiquitin-protein ligase involved in Golgi membrane fusion and regulation of small GTPases. Acts as a regulator of Golgi membrane dynamics during the cell cycle: recruited to Golgi membrane by Rab proteins and regulates postmitotic Golgi membrane fusion. Acts by mediating ubiquitination during mitotic Golgi disassembly, ubiquitination serving as a signal for Golgi reassembly later, after cell division. Specifically interacts with GTP-bound RAC1, mediating ubiquitination and subsequent degradation of active RAC1, thereby playing a role in host defense against pathogens. May also act as a transcription regulator via its interaction with RARB.Specifications
| HACE1 | |
| Monoclonal | |
| Unconjugated | |
| HACE1 | |
| SPPRS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57531 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6H8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8IYU2 | |
| HACE1 | |
| Full length human recombit protein of human HACE1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.