missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADH Antibody (OTI3D12), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70875V
This item is not returnable.
View return policy
Description
HADH Monoclonal specifically detects HADH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HADH | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HADH | |
| Recombinant protein fragment corresponding to amino acids 57-314 of human HADH (NP_005318) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI3D12 | |
| Western Blot, Immunohistochemistry | |
| EC 1.1.1, EC 1.1.1.35, HADH1, HADHSChydroxyacyl-Coenzyme A dehydrogenase, HCDH, hydroxyacyl-CoA dehydrogenase, L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain, Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase, MGC8392, mitochondrial, MSCHAD, SCHADHHF4 | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 3033.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction