Learn More
HADH Mouse anti-Human, Clone: UMAB151, lyophilized, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM800048CF
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.Specifications
| HADH | |
| Monoclonal | |
| Unconjugated | |
| HADH | |
| HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3033 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Peptide Array, Western Blot | |
| UMAB151 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q16836 | |
| HADH | |
| Human recombit protein fragment corresponding to amino acids 57-314 of human HADH produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.