missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64551C
This item is not returnable.
View return policy
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Haemophilus Influenzae B | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. | |
| Store at 4°C in the dark. |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1079/457 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| H. influenza B | |
| Native, NCTC 7279 | |
| Primary | |
| Bacteria | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction