missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/83), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64552AF594
This item is not returnable.
View return policy
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Haemophilus Influenzae B | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-64552 is reactive with Heamophilus Influenzae B. NB100-64552 is reactive with isolates fixed by phenol, formalin and boiling. | |
| Store at 4°C in the dark. |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1079/83 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| H. influenza B | |
| Native, NCTC 7279 | |
| Primary | |
| Bacteria | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction