missing translation for 'onlineSavingsMsg'
Learn More

Haemophilus Influenzae B Antibody (1079/83), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NB100-64552CL1

Product Code. 30156908

  • € 1020.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Haemophilus Influenzae B Monoclonal antibody specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Haemophilus Influenzae B
Monoclonal
CoraFluor 1
H. influenza B
Native Haemophilus influenzae B
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG2b
ELISA, Immunocytochemistry/Immunofluorescence
1079/83
PBS
Mouse
Protein A purified
RUO
Bacteria
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.