missing translation for 'onlineSavingsMsg'
Learn More

Hafnium, plasma standard solution, Specpure™, Hf 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7049293
40.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00011032
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00011032

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration HfCl2O in 5% HCl
Quantity 50 mL
UN Number UN1789
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Hafnium plasma standard solution, Hf 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'