missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAGH Monoclonal Antibody (611CT23.6.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537610
This item is not returnable.
View return policy
Description
HAGH Monoclonal Antibody for Western Blot
The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
Specifications
| HAGH | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.09% sodium azide; pH 7.4 | |
| Q16775 | |
| HAGH | |
| KLH conjugated synthetic peptide between 279-308 amino acids from the C-terminal region of human HAGH. | |
| RUO | |
| 3029 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 611CT23.6.1 | |
| Unconjugated | |
| HAGH | |
| BC019817; GLO2; Glo-2; Glx II; GLX2; GLXII; Glyoxalase II; glyoxylase 2; HAGH; HAGH1; hydroxyacyl glutathione hydrolase; hydroxyacylglutathione hydrolase; hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; round spermatid protein RSP29; Rsp29 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction