missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAGHL Antibody (OTI3H2), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72283PECY55
This item is not returnable.
View return policy
Description
HAGHL Monoclonal specifically detects HAGHL in Human samples. It is validated for Flow Cytometry.
Specifications
| HAGHL | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| HAGHL | |
| Full length recombinant protein of human HAGHL (NP_115680) produced in HEK293T cells. | |
| 0.1 mL | |
| 84264.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI3H2 | |
| Flow Cytometry | |
| EC 3.1.2.-, GLO2-like/ RJD12, hydroxyacyl glutathione hydrolase-like, hydroxyacylglutathione hydrolase-like, hydroxyacylglutathione hydrolase-like protein, MGC2605 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction