missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hairy (Drosophila) Antibody (3/30), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50417AF488
This item is not returnable.
View return policy
Description
Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.
Specifications
| Hairy (Drosophila) | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Hairy gene product | |
| 0.1 mL | |
| Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
| Drosophila | |
| Purified |
| Western Blot | |
| 43189 | |
| Western Blot | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction