Learn More
Zeiss™ Halogenogen Reflector Bulbs
Product Code. 10741971
Change view
Click to view available options
Wattage:
250 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage Unit Size
10741971 250 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10741971 Supplier Zeiss™ Supplier No. 0000000300271
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
For use with Carl Zeiss Microscopes
  • Bulb
  • Halogen Reflector (D)
  • 24V
  • 250W
Specifications
Light Source Halogen
Voltage 24 V
Wattage 250 W
Product Type Halogen Reflector Bulb