missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol Decanoate, Mikromol™

Product Code. 30052403
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052403 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052403 Supplier Mikromol Supplier No. MM1276.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Haloperidol Decanoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Haloperidol decanoate
CAS 74050-97-8
Analyte or Component Names Haloperidol decanoate
Shipping Condition Room Temperature
Molecular Formula C31 H41 Cl F N O3
Certifications/Compliance ISO 17034
Synonym Haloperidol decanoate, 4-(4-Chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]piperidin-4-yl decanoate
InChI Formula InChI=1S/C31H41ClFNO3/c1-2-3-4-5-6-7-8-11-30(36)37-31(26-14-16-27(32)17-15-26)20-23-34(24-21-31)22-9-10-29(35)25-12-18-28(33)19-13-25/h12-19H,2-11,20-24H2,1H3
SMILES CCCCCCCCCC(=O)OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3
IUPAC Name [4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]piperidin-4-yl] decanoate
Molecular Weight (g/mol) 530.11
Quantity 250 mg
Formula Weight 529.2759 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.