Learn More
Hamilton™ 700 Series Microliter™ Manual Injection Syringe for JADE Equipd Agilent Autosamp-REV
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Description
MICROLITER syringe with a stainless steel plunger that is hand-fitted to the glass barrel, designed for liquid sampling applications. Syringe has a cemented (N) termination. Available in 10μL volume.
Syringe Termination
- Cemented Needle (N)- For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
- Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauge: 23s
Specifications
Specifications
| Needle Gauge | 23s G |
| Length (English) | 2.5 in. |
| Length (Metric) | 65 mm |
| Needle Point Number | AS |
| Volume (Metric) | 10 μL |