Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Click to view available options
For Use With (Equipment):
Hamilton Syringe Models 702-750, 7000 and 1700 Series
Hamilton Syringe Models 75 and 701 Series
Hamilton Syringe Models 800 and 1800 Series
Unit Size:
Each
Description
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Spécification
Spécification
| Product Type | Reproducibility Adapter |
| Quantity | Ea. |
| For Use With (Equipment) | Hamilton Syringe Models 800 and 1800 Series |