Learn More
Hamilton™ HxSil C18 Guard Cartridge Kit
Click to view available options
:
C18 Guard Cartridge Kit, Stainless Steel
Description
- Guard columns remove particulate contaminants and highly adsorptive compounds from samples
- Modular and cartridge replacement is easy and tool free
- Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
- Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except, concentrated nitric acid, sulfuric acid, tetrahydrofuran or dimethylsulfoxide.
Specifications
Specifications
| Length (Metric) | 20 mm |
| Stationary Phase | C18 |
| Material | Stainless Steel |