Learn More
Hamilton™ Metal Hub Side Port Needles (Luer Lock)
Click to view available options
Needle Gauge:
18 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
Unit Size:
Pack of 6
Description
Metal Hub Compatible Syringe Terminations
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
Six needles per pack.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nickel-plated Brass (Hub), Stainless Steel |
| Needle Gauge | 26 G |
| Diameter (English) Inner | 0.01 in. |
| Diameter (English) Outer | 0.02 in. |
| Diameter (Metric) Inner | 0.26 mm |
| Diameter (Metric) Outer | 0.46 mm |
| Length (English) Needle | 2 in. |
| Product Type | Replacement Needle |
| Needle Point Style | Cone with Side Port |
| Show More |