Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Brand: Hamilton™ 240134
Additional Details : Weight : 1.36080kg
Description
- For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
FEP | |
1.29 mL | |
1/4-28 Tubing threads | |
1372 mm |
Gauge 18, Dispense Tubing | |
Microlab™ 600 diluter/dispenser system | |
54.01 in. |