Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Brand: Hamilton™ 1174-02
Additional Details : Weight : 0.20000kg
Description
- For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
FEP | |
0.94 μL/mm | |
1/4-28 Tubing threads |
Gauge 18, Fill Tubing | |
Microlab™ 600 diluter/dispenser system |