Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Brand:  Hamilton™ 1172-02
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15867074
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
Specifications
FEP
3.75 μL/mm
1/4-28 Tubing threads
Gauge 12, Fill Tubing
Microlab™ 600 diluter/dispenser system