Learn More
Hamilton™ Microliter™ and Gastight™ Syringes: Model 75 for Agilent 7673, 7683, 7693 and 6850 ALS GC
Product Code. 11547844
Change view
Click to view available options
Needle Gauge:
23 to 26s G
23s G
26s G
Model:
75 N (Cemented Needle)
75 RN (Removable Needle)
Needle Point Number:
2
AS
Quantity:
6 Pk.
Ea.
Unit Size:
Each
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Model Needle Point Number Quantity unitSize
11547844 23 to 26s G 75 N (Cemented Needle) AS Ea. Each
10268910 23 to 26s G 75 RN (Removable Needle) AS Ea. Each
10196920 23 to 26s G 75 N (Cemented Needle) AS 6 Pk. Pack of 6
10116880 23s G 75 RN (Removable Needle) AS Ea. Each
10190261 23s G 75 N (Cemented Needle) AS Ea. Each
10702392 26s G 75 N (Cemented Needle) AS Ea. Each
10321001 26s G 75 N (Cemented Needle) AS 6 Pk. Pack of 6
10780652 23s G 75 N (Cemented Needle) AS 6 Pk. Pack of 6
10770034 23s G 75 N (Cemented Needle) 2 Ea. Each
10522378 26s G 75 N (Cemented Needle) 2 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11547844 Supplier Hamilton™ Supplier No. 87993
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
Designed to work with Agilent 7673, 7683, 7693 and 6850 Automatic Liquid Sampler (ALS) gas chromatography (GC) autosamplers
Microliter syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling.
Syringe Terminations
  • Cemented Needle (N) - For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
  • Removable Needle (RN) - The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Styles
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 23s or 26s
  • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 23s, 26s, 23s-26s
Specifications
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Needle Gauge 23 to 26s G
Diameter (English) Inner 0.014 in.
Diameter (English) Inner Needle 0.0133 to 0.005 in.
Diameter (Metric) Inner Needle 0.64 to 0.13 mm
Diameter (English) Outer Needle 0.0283 to 0.0187 in.
Diameter (English) Outer 0.26 in.
Diameter (Metric) Inner 0.343 mm
Diameter (Metric) Outer 6.6 mm
Diameter (Metric) Outer Needle 0.72 to 0.47 mm
Length (English) 1.7 in.
Length (English) Needle 1.71 in.
Length (Metric) 43 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 75 N (Cemented Needle)
Needle Point Number AS
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 5 μL
Needle Point Style Cone
For Use With (Equipment) Agilent 7673, 7683, 6850 Automatic Liquid Sampler, Gas Chromatography Autosamplers
Connection Type Cemented