Learn More
Hamilton™ PRP™Series Guard Cartridge Replacement Cartridges
Product Code. 10686591
Click to view available options
Sorbent:
C18
PRP-1
PRP-X100
PRP-X200
Diameter (Metric) Inner:
2.0 mm
4 mm
For Use With (Equipment):
Hypersil™ HPLC Column
Organic Acid Analysis Column
Unit Size:
Pack of 5
Product Code. 10686591
Brand: Hamilton™ 79446
Only null left
Add to basket
Add to basket
Replacement Cartridges for PRP-1, PRP-3, PRP-X100, PRP-X200, PRP-X300, PRP-X400
Stainless steel or PEEK replacement cartridges.
Specifications
pH 1 to 13
Particle Size 10 μm
Length (Metric) 25 mm
Max. Pressure 5000 psi
Diameter (Metric) Inner 4 mm
For Use With (Equipment) Hypersil™ HPLC Column
Material 316 Stainless Steel
Sorbent PRP-X100