missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd69, Hamster, SPRD, Clone: H1.2F3, Abnova™
Hamster monoclonal antibody raised against native Cd69.
Brand: Abnova MAB5746.100ug
This item is not returnable.
View return policy
Specifications
| Cd69 | |
| Monoclonal | |
| SPRD | |
| Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd69 | |
| Cd69 | |
| Native purified Cd69 from Y245 mouse dendritic epidermal T cell line. | |
| RUO | |
| 12515 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| H1.2F3 | |
| Hamster monoclonal antibody raised against native Cd69. | |
| In PBS (0.09% sodium azide) | |
| 5830438K24Rik/AI452015/AIM/VEA | |
| Hamster | |
| 100 μg | |
| Primary | |
| Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 KDa (unreduced). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction