Learn More
CD30 Hamster anti-Mouse, Brilliant Violet 421, Clone: mCD30.1 (also known as 2SH12-5F-2D), BD Optibuild™
Hamster Monoclonal Antibody
Brand: BD Biosciences 740031
Description
The mCD30.1 monoclonal antibody specifically recognizes CD30. CD30 is also known as Tumor necrosis factor receptor superfamily, member 8 (Tnfrsf8). The CD30 molecule is predominantly expressed by activated T lymphocytes, with its expression peaking at day 4-5 on spleen cells activated with plate-bound anti-CD3e antibody. The mCD30.1 antibody reacts with a majority of CD8+ T cells, as well as some CD4+ T cells in these cultures. Expression of CD30 on activated T lymphocytes is regulated by CD28 and cytokines. Its TNF-superfamily ligand, CD30L or CD153, is also expressed on activated T lymphocytes. By northern blot analysis, mouse Cd30 mRNA is detected in the thymus and in 72-hour pokeweed mitogen- and Con A-activated spleen cells, but not in the lung, brain, kidney, liver, bone marrow, unactivated spleen, or 72-hour LPS-activated splenocytes.1 It has also been reported that CD30 is expressed on naive B lymphocytes, it is not detectable after activation, and it starts to return after 72 hours following activation. Reports suggest that signaling through the CD30 molecule may be important in cytokine production by CD8+ CTL lines and may play a role in the regulation of Th1 and Th2 cytokine secretion by CD4+ and CD8+ T cells. It has also been proposed that CD30 plays an important role in the negative selection of thymocytes and protects against autoimmunity. Members of the TNFR family and their ligands are involved in the induction of diverse biological responses in lymphocytes, including differentiation, proliferation, and cellular death. In humans, CD30 was initially identified in Hodgkin and Reed-Sternberg cells in Hodgkin’s disease patients and subsequently was found on neoplastic cells of certain types of non-Hodgkin’s lymphomas.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
| CD30 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd30; Tnfrsf8; Ki; Ki--1; CD30L Receptor | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| mCD30.1 (also known as 2SH12-5F-2D) | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CD30-IgG1 fusion protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only