missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamster IgG1, λ1 Isotype Control, BV711, Clone: G235-2356, BD
Brand: BD Biosciences 563049
Additional Details : Weight : 0.01000kg
Description
Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, λ 1
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
G235-2356 | |
Brilliant Violet 711 | |
Trinitrophenol-KLH | |
50 μg | |
Isotype Control (Primary) | |
IgG1 λ1 |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |