Learn More
HAND1 Antibody (OTI1G10), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71459AF700
Product Code. 18105782
  • € 796.00 / 0.10mL
Description
HAND1 Monoclonal specifically detects HAND1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HAND1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
HAND1
Full length recombinant protein of human HAND1 (NP_004812) produced in E.coli.
0.1 mL
9421.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1G10
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, EHAND, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2b