Learn More
HAND1 Mouse anti-Human, Clone: OTI1G10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502671
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.Specifications
| HAND1 | |
| Monoclonal | |
| Unconjugated | |
| HAND1 | |
| Hxt, Thing1, bHLHa27, eHand | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9421 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1G10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O96004 | |
| HAND1 | |
| Full length human recombit protein of human HAND1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.