missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAND2, Mouse anti-Human, Clone: 1C7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009464-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: LSKIKTLRLATSYIAYLMDLLAKDDQNGEAEAFKAEIKKTDVKEEKRKKELNEILKSTVSSNDKKTKGRTGWPQHVWALELKSpecifications
| HAND2 | |
| Monoclonal | |
| Unconjugated | |
| heart and neural crest derivatives expressed 2 | |
| DHAND2/FLJ16260/Hed/MGC125303/MGC125304/Thing2/bHLHa26/dHand | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1C7 | |
| In 1x PBS, pH 7.4 | |
| NM_021973 | |
| HAND2 | |
| HAND2 (NP_068808.1, 135 a.a. ∼ 216 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9464 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction