missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hantavirus NP Monoclonal Antibody (J41L)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA518209

Product Code. 15332687

  • € 476.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific for the nucleocapsid protein (N) of all Hantaviruses tested: Hantaan, Dobrava, Seoul and Saaremaa. Purity is >90%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hantavirus NP
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
ELISA, Immunocytochemistry
J41L
Unconjugated
HV; HV nucleoprotein
Hantavirus recombinant protein.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.