missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAO1 Mouse anti-Human, Clone: UMAB117, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM570082
This item is not returnable.
View return policy
Description
Has 2-hydroxyacid oxidase activity. Most active on the 2-carbon substrate glycolate, but is also active on 2-hydroxy fatty acids, with high activity towards 2-hydroxy palmitate and 2-hydroxy octanoate.Specifications
| HAO1 | |
| Monoclonal | |
| 0.5-1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJM8 | |
| HAO1 | |
| Full length human recombit protein of human HAO1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| UMAB117 | |
| Unconjugated | |
| HAO1 | |
| GOX, GOX1, HAOX1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction