missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAP1 Antibody (1B6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-74569FR
This item is not returnable.
View return policy
Description
HAP1 Monoclonal antibody specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| HAP1 | |
| Monoclonal | |
| DyLight 680 | |
| HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1 | |
| Fusion protein from rat HAP1. [UniProt# P54256] | |
| 0.1 mL | |
| Autophagy, Neuroscience | |
| 9001 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 1B6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction