missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52692UV
This item is not returnable.
View return policy
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal antibody specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF
Specifications
| hapten 4-hydroxy-3-nitrophenyl acetyl (NP) | |
| Monoclonal | |
| DyLight 350 | |
| (4-Hydroxy-3-nitrophenyl)acetyl, 2-(4-Hydroxy-3-nitrophenyl)acetate, 2-(4-Hydroxy-3-nitrophenyl)acetic acid, 3-nitro-4-Hydroxyphenylacetic acid, 4-Hydroxy-3-nitrophenyl acetyl | |
| hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 λ |
| Western Blot, Flow Cytometry, Control, CyTOF | |
| B1-8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction