missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric Recombinant Rat Monoclonal Antibody (B1-8)

Rat Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551700
This item is not returnable.
View return policy
Description
Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).
Hapten 4-hydroxy-3-nitrophenyl acetyl NP is a small molecule that consists of a hapten, which is the 4-hydroxy-3-nitrophenyl acetyl group, linked to a carrier protein, which is NP or nitrophenol. The carrier protein is an important component of this molecule as it helps to elicit an immune response by presenting the hapten portion of the molecule to the immune system. NP is commonly used as a carrier in immunology research because it is a small, soluble molecule that is easily conjugated with haptens. When the hapten binds to specific antibodies, the carrier protein helps to stimulate an immune response and activate B cells, which can lead to the production of specific antibodies against the hapten.
Specifications
| Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| 4-hydroxy-3-nitrophenyl acetyl (NP) - conjugated to NP12-chicken gammaglobulin (CG). | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Functional Assay | |
| B1-8 | |
| Unconjugated | |
| Rat | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction