Learn More
Haptoglobin Antibody (5A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26220-100ul
 View more versions of this product
Product Code. 30025659
  • € 533.00 / 100µL
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Haptoglobin
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
5A5
ELISA, Immunohistochemistry 1:50-1:200
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin
A synthesized peptide derived from Human Haptoglobin [UniProt P00738]
100 μL
Cardiovascular Biology
3240
Store at -20 to -70C. Avoid freeze-thaw cycles.