missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Antibody (HP/3838), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26953CL1
This item is not returnable.
View return policy
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Haptoglobin | |
| Monoclonal | |
| CoraFluor 1 | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Recombinant fragment (around aa100-250) of human Haptoglobin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 3240 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| HP/3838 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction