missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Antibody (OTI4H5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70925AF350
This item is not returnable.
View return policy
Description
Haptoglobin Monoclonal specifically detects Haptoglobin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Haptoglobin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HP | |
| Full length recombinant protein of human Haptoglobin (NP_005134) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4H5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Mouse | |
| 0.1 mL | |
| 3240 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction