missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Haptoglobin Recombinant Rabbit Monoclonal Antibody (7C1N2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535716
This item is not returnable.
View return policy
Description
Immunogen sequence: DQCIRHYEGS TVPEKKTPKS PVGVQPILNE HTFCAGMSKY QEDTCYGDAG SAFAVHDLEE DTWYATGILS FDKSCAVAEY GVYVKVTSIQ DWVQKTIAEN.
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.
Specifications
| Haptoglobin | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00738, P06866, Q61646 | |
| HP | |
| A synthetic peptide corresponding to a sequence within amino acids 307-406 of human Haptoglobin (HP) (P00738). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7C1N2 | |
| Unconjugated | |
| HP | |
| alpha polypeptide; Ba1-647; beta polypeptide; binding peptide; Binding peptide (BP); BP; haptoglobin; haptoglobin alpha 1S; Haptoglobin alpha chain; Haptoglobin alpha chain / alpha(1S) beta; haptoglobin alpha(1S)-beta; Haptoglobin alpha(2FS) beta; haptoglobin alpha(2FS)-beta; Haptoglobin beta chain; haptoglobin, alpha polypeptide; haptoglobin, beta polypeptide; haptoglobin; haptoglobin-like; haptoglobin-like; haptoglobin-related protein; Hp; HP-1; HP2 ALPHA2; HP2ALPHA2; HP2-ALPHA-2; HPA1S; HPR; HPT; Liver regeneration-related protein LRRG173; LOC100067869; LOC479668; MGC111141; MGC133752; preHP2; zonulin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15439, 24464, 3240 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction