Learn More
WNT3 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801734S
Encompass_Preferred
 View more versions of this product
Product Code. 17520143
  • € 242.00 / 30µL
Description
WNT3 Monoclonal antibody specifically detects WNT3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
WNT3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00443
Mouse
Affinity chromatography
RUO
5286
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1C5
Unconjugated
PIK3C2A
INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein
Human recombinant protein fragment corresponding to amino acids 230-560 of human PIK3C2A produced in E.coli.