Learn More
Hastelloy C™ gauze, 40 mesh woven from 0.19mm (0.0075 in.) dia. wire, Thermo Scientific Chemicals
€ 33.25 - € 321.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427285
View Documents Promotion Details
Thermo Scientific Alfa Aesar
046690.FL
75 x 75 mm
€ 33.25
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
15437285
View Documents Promotion Details
Thermo Scientific Alfa Aesar
046690.GT
150 x 150 mm
€ 97.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
15447285
View Documents Promotion Details
Thermo Scientific Alfa Aesar
046690.HB
300 x 300 mm
€ 321.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-05-2024
to see stock.
Description
Description
Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.