missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAUS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534874
This item is not returnable.
View return policy
Description
Predicted reactivity: Mouse (84%), Rat (85%), Pig (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications
| HAUS1 | |
| Polyclonal | |
| Unconjugated | |
| HAUS1 | |
| BC024400; CCDC5; coiled-coil domain containing 5; coiled-coil domain containing 5 (spindle associated); coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin like complex subunit 1; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HAUS1; HEIC; HEI-C; HsT1461; spindle associated | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115106 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.43 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96CS2 | |
| HAUS1 | |
| Full length human HAUS1 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction