missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAUS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556264
This item is not returnable.
View return policy
Description
Immunogen sequence: RCLTLLQRLL QEHRLKTQSE LDRINAQYLE VKCGAMILKL RMEELKILSD TYTVEKVEVH RLIRDRLEGA IHLQEQDMEN SRQVLNSYEV LGEEFDRLVK EYTVLKQATE NKRWALQEFS KVYR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 83%.
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintenance of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.
Specifications
| HAUS4 | |
| Polyclonal | |
| Unconjugated | |
| Haus4 | |
| 9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS augmin like complex subunit 4; HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; HAUS4; homologous to Augmin subunits; RGD1305288 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H6D7 | |
| Haus4 | |
| Recombinant protein corresponding to Human HAUS4. Recombinant protein control fragment (Product #RP-102183). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction