Learn More
HAUS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20104-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintece of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.Specifications
| HAUS4 | |
| Polyclonal | |
| Unconjugated | |
| HAUS4 | |
| C14orf94, HAUS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219072, 305882, 54930 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BFT2, Q9H6D7 | |
| Haus4 | |
| HAUS4 Fusion Protein Ag13717 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |