Learn More
HAUS7 Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507056
This item is not returnable.
View return policy
Description
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.Specifications
| HAUS7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99871 | |
| HAUS7 | |
| Full length human recombit protein of human HAUS7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| HAUS7 | |
| UCHL5IP, UIP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55559 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |