missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAUS7 Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507056S
This item is not returnable.
View return policy
Description
HAUS7 Monoclonal antibody specifically detects HAUS7 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| HAUS7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99871 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55559 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| HAUS7 | |
| 1110020L19Rik; 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin like complex subunit 7; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; Haus7; RGD1562991; UCH37-interacting protein 1; UCHL5 interacting protein; UCHL5-interacting protein; UCHL5IP; UIP1; X-linked protein STS1769 | |
| Full length human recombinant protein of human HAUS7 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction