missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAUS7 Recombinant Rabbit Monoclonal Antibody (10L5Y8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555588
This item is not returnable.
View return policy
Description
Immunogen Sequence: LMEHFEDTRE KNEALLGELF SSPHLQMLLN PECDPWPLDM QPLLNKQSDD WQWASASAKS EEEEKLAELA RQLQESAAKL HALRTEYFAQ HEQGAAAGAA DISTLDQKLR LVTSDFHQLI LAFLQVYDDE LGECCQ.
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
Specifications
| HAUS7 | |
| Recombinant Monoclonal | |
| 1.817 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99871 | |
| HAUS7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-285 of human UCHL5IP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 10L5Y8 | |
| Unconjugated | |
| HAUS7 | |
| 1110020L19Rik; 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin like complex subunit 7; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; Haus7; RGD1562991; UCH37-interacting protein 1; UCHL5 interacting protein; UCHL5-interacting protein; UCHL5IP; UIP1; X-linked protein STS1769 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55559 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction