missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAVCR2 Monoclonal Antibody (OTI13A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812578
This item is not returnable.
View return policy
Description
TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.Specifications
| HAVCR2 | |
| Monoclonal | |
| Unconjugated | |
| HAVCR2 | |
| CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84868 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI13A8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TDQ0 | |
| HAVCR2 | |
| Full length human recombit protein of human HAVCR2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction