missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAVCR2 Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812558S
This item is not returnable.
View return policy
Description
HAVCR2 Monoclonal antibody specifically detects HAVCR2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HAVCR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TDQ0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84868 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1G9 | |
| Unconjugated | |
| Havcr2 | |
| CD366; FLJ14428; Havcr2; HAVcr-2; Hepatitis A virus cellular receptor 2; hepatitis A virus cellular receptor 2 homolog; kidney injury molecule-3; KIM-3; sCD366; soluble CD366; soluble TIM 3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain containing 3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; Tim3; TIM-3; TIMD3; TIMD-3 | |
| Full length human recombinant protein of human HAVCR2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction