missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAX-1 Antibody (3C5), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30169MFV500
This item is not returnable.
View return policy
Description
HAX-1 Monoclonal antibody specifically detects HAX-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| HAX-1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 3C5 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3 | |
| Recombinant human HAX-1 (1-279aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis | |
| 10456 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction