Learn More
New Pig™ HazMat Spill Kits
Click to view available options
Capacity (Metric):
13.62 L
34.068 L
79.49 L
For Use With (Application):
Acids, Bases, Unknowns
Acids, Bases, Unknowns Fluid Absorption
Oils, Coolants, Solvents, Water
Unit Size:
Each
Description
- Feature PIG absorbents proven for rapid response
- Socks stop spreading spills; PIG HazMat Pads absorb quickly
- Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
- Temporary disposal bags help make cleanup easier
- Bright yellow bag and large, color-coded label make the kit easy to locate
- Strap for easy carrying and hanging
- Compact and easy to store
Specifications
Specifications
| Capacity (English) | 9 gal. |
| Capacity (Metric) | 34.068 L |
| Material | Vinyl-laminated Nylon, Polypropylene |
| Type | Spill Response Bag |
| For Use With (Application) | Acids, Bases, Unknowns |
| Includes | 4 external dia. 3 in., 46 in. L PIG&trade, HazMat Chemical Absorbent Sock, 28 L x W: 20 x 15 in. PIG&trade, HazMat Mat Pad, 5 W x H: 18 x 30 in. Polyethylene Disposal Bags, 1 Instructions |
| Weight (English) | 11.9 lb. |
| Weight (Metric) | 5.4 kg |